5-(Trimethylsilyl)-1,3-cyclopentadiene ≥97% (mixture of isomers)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Trimethylsilylcyclopentadiene (mixture of isomers), (2,4-Cyclopentadien-1-yl)trimethylsilane

14-7950-25G 14-7950-1G 14-7950-5G
100197-382EA 897.35 USD
100197-382 100208-314 100202-932
5-(Trimethylsilyl)-1,3-cyclopentadiene ≥97% (mixture of isomers)
5-(Trimethylsilyl)-1,3-cyclopentadiene

Metallocenes, Derivatives & Cp Precursors

Formula: C₈H₁₄Si
MW: 138.29 g/mol
Boiling Pt: 138…140 °C
Density: 0.833
Flash Pt: 85 °F
MDL Number: MFCD00010666
CAS Number: 3559-74-8
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR