Silica gel, pore_large

Supplier: STREM CHEMICALS INC MS

14-7420-250G 14-7420-1000G
100198-580EA 59.45 USD
100198-580 100199-760
Silica gel, pore_large
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR