Silica gel, large pore

Supplier: STREM CHEMICALS INC MS

14-7400-250G 14-7400-1000G
100204-910EA 42.46 USD
100204-910 100209-658
Silica gel, large pore
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR