Silicon(IV) phthalocyanine dichloride ≥85%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
14-2000-5G
14-2000-1G
100208-312EA
835.08
USD
100208-312
100207-978
Silicon(IV) phthalocyanine dichloride ≥85%
Silicon(IV) phthalocyanine dichloride
Formula:
C₃₂H₁₆Cl₂N₈Si Melting Pt: 430 °C (subl.) |
MDL Number:
MFCD00010723 CAS Number: 19333-10-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...