Dimethylbis(indenyl)silane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

14-1950-25G 14-1950-5G
100206-082EA 1729.6 USD
100206-082 100198-118
Dimethylbis(indenyl)silane ≥98%
Dimethylbis(indenyl)silane

Metallocenes, Derivatives & Cp Precursors

MDL Number: MFCD00674015
CAS Number: 136946-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR