tert-Butyldimethylsilyl trifluoromethanesulfonate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: t-Butyldimethylsilyl trifluoromethanesulphonate

14-1800-5G 14-1800-1G 14-1800-25G
100199-758EA 215.14 USD
100199-758 100204-906 100202-634
tert-Butyldimethylsilyl trifluoromethanesulfonate ≥98%
tert-Butyldimethylsilyl trifluoromethanesulphonate
Formula: C₇H₁₅F₃O₃SSi
MW: 264.34 g/mol
Density: 1.151
Flash Pt: 98 °F
MDL Number: MFCD00000405
CAS Number: 69739-34-0
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR