(1S,2S)-(+)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]aluminum(III) chloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S,S)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminoaluminum chloride

13-5801-5G 13-5801-1G
100204-904EA 1065.78 USD
100204-904 100204-902
(1S,2S)-(+)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]aluminum(III) chloride ≥98%
(1S,2S)-(+)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]aluminum(III) chloride
Formula: C₃₆H₅₂AlClN₂O₂
Melting Pt: >350 °C
MDL Number: MFCD02684548
CAS Number: 307926-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR