Dimethylaluminum i-propoxide ≥98% (99.99+%-Al), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

13-1600-5G 13-1600-1G
101324-536EA 687.88 USD
101324-536 101324-534
Dimethylaluminum i-propoxide ≥98% (99.99+%-Al), PURATREM
Dimethylaluminum i-propoxide

Alkoxide, Metal Alkoxides, Metal Alkyls, Volatile Precursors for CVD

Density: 0.824
MDL Number: MFCD07369026
CAS Number: 6063-89-4
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR