Aluminum hexafluoro-2,4-pentanedionate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
13-0200-25G
13-0200-5G
100210-610EA
1346.03
USD
100210-610
100198-114
Aluminum hexafluoro-2,4-pentanedionate ≥98%
Aluminum hexafluoro-2,4-pentanedionate
Metal Beta-diketonates, Volatile Precursors for CVD
Formula:
C₁₅H₃AlF₁₈O₆ Boiling Pt: 170 °C/ 50 °C (0.1 mmHg) (dec./ subl.) Melting Pt: 70…73 °C |
MDL Number:
MFCD00050051 CAS Number: 15306-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...