Magnesium meso-tetraphenylporphine hydrate

Supplier: STREM CHEMICALS INC MS

12-2000-1G 12-2000-5G
100209-642EA 380.74 USD
100209-642 100204-498
Magnesium meso-tetraphenylporphine hydrate
Magnesium meso-tetraphenylporphine hydrate

Porphine

Formula: (C₄₄H₂₈N₄)Mg·xH₂O
MDL Number: MFCD00150433
CAS Number: 14640-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR