Aluminium magnesium isopropoxide ≥99.99% (Mg basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Magnesium aluminum i-propoxide

12-1100-10G 12-1100-50G
100197-360EA 523.69 USD
100197-360 100207-204
Aluminium magnesium isopropoxide ≥99.99% (Mg basis), PURATREM
Aluminium magnesium isopropoxide

Alkoxide

Formula: C₂₄H₅₆Al₂MgO₈
MW: 550.965 g/mol
Boiling Pt: 110 °C (5 mmHg)
Melting Pt: 40 °C
MDL Number: MFCD00070426
CAS Number: 69207-83-6
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR