Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium(II) dihydrate ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium dihydrate, Mg(TMHD)2, Magnesium-DPM dihydrate

12-1000-5G 12-1000-1G 12-1000-25G
100210-980EA 341.11 USD
100210-980 100210-976 100210-978
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium(II) dihydrate ≥98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')magnesium dihydrate

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₂₂H₃₈MgO₄·2H₂O
MW: 426.87 g/mol
Boiling Pt: 290 °C/ 150 °C (0.05 mmHg) (dec./ subl.)
Melting Pt: 135…150 °C (-H₂O)
MDL Number: MFCD06200256
CAS Number: 625832-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR