Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium, Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')magnesium, [Mg(TMHD)2]

12-0900-1G 12-0900-5G
100210-972EA 103.32 USD
100210-972 100210-974
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium, anhydrous ≥98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₂₂H₃₈MgO₄
MW: 390.85 g/mol
MDL Number: MFCD00070425
CAS Number: 21361-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR