Ethylmagnesium chloride 2M in ether

Supplier: STREM CHEMICALS INC MS
Danger

12-0800-1MOL 12-0800-200MMOL
100197-060EA 331.2 USD
100197-060 100206-284
Ethylmagnesium chloride 2M in ether
Ethylmagnesium chloride

Grignard

Formula: C₂H₅ClMg
MDL Number: MFCD00000472
CAS Number: 2386-64-3
UN: 3399
ADR: 4.3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR