Bis(n-propylcyclopentadienyl)magnesium ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

12-0550-5G 12-0550-1G
100202-920EA 830.83 USD
100202-920 100206-282
Bis(n-propylcyclopentadienyl)magnesium ≥98%
Bis(n-propylcyclopentadienyl)magnesium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₆H₂₂Mg
Density: 0,94
MDL Number: MFCD00151384
CAS Number: 114504-74-4
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR