Bis(ethylcyclopentadienyl)magnesium ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

12-0510-1G 12-0510-5G 12-0510-25G
100203-952EA 141.54 USD
100203-952 100203-956 100203-954
Bis(ethylcyclopentadienyl)magnesium ≥98%
Bis(ethylcyclopentadienyl)magnesium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₄H₁₈Mg
Density: 1,912
MDL Number: MFCD00151375
CAS Number: 114460-02-5
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR