Bis(cyclopentadienyl)magnesium ≥99.9% (Mg basis)

Supplier: STREM CHEMICALS INC MS
Danger

12-0500-25G 12-0500-1G 12-0500-5G
100206-280EA 1554.09 USD
100206-280 100209-638 100209-640
Bis(cyclopentadienyl)magnesium ≥99.9% (Mg basis)
Bis(cyclopentadienyl)magnesium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₀H₁₀Mg
MW: 154.49 g/mol
Boiling Pt: 290 °C/ 160 °C (0,1 mmHg) (dec./ subl.)
Melting Pt: 176 °C
MDL Number: MFCD00134487
CAS Number: 1284-72-6
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR