Cacodylic acid sodium salt trihydrate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
11-1000-25G
11-1000-100G
100209-634EA
124.55
USD
100209-634
100198-574
Cacodylic acid sodium salt trihydrate ≥98%
Cacodylic acid sodium salt trihydrate
Formula:
(CH₃)₂As(O)ONa·3H₂O |
MDL Number:
MFCD00149079 CAS Number: 6131-99-3 UN: 1688 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...