Perfluorodecahydronaphthalene (mixture of cis and trans isomers) ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

09-5960-25G 09-5960-5G
100203-934EA 104.74 USD
100203-934 100204-874
Perfluorodecahydronaphthalene (mixture of cis and trans isomers) ≥95%
Perfluorodecahydronaphthalene (mixture of cis and trans isomers)

Organo Fluorine

Formula: C₁₀F₁₈
Boiling Pt: 141 °C
Melting Pt: –10 °C
Density: 1,908
Flash Pt: 105 °F
MDL Number: MFCD00010626
CAS Number: 306-94-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR