2,2,3,3,4,4,4-Heptafluorobutylamine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

09-4396-5G 09-4396-1G
100209-602EA 484.06 USD
100209-602 100209-600
2,2,3,3,4,4,4-Heptafluorobutylamine ≥97%
2,2,3,3,4,4,4-Heptafluorobutylamine

Organo Fluorine

Formula: C₄H₄F₇N
MW: 199.07 g/mol
Boiling Pt: 70…71 °C
Density: 1,493
MDL Number: MFCD00014817
CAS Number: 374-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR