Trimethyloxonium tetrafluoroborate ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
08-2500-5G
08-2500-25G
100195-994EA
110.4
USD
100195-994
100207-948
Trimethyloxonium tetrafluoroborate ≥95%
Trimethyloxonium tetrafluoroborate
Formula:
C₃H₉BF₄O MW: 147.91 g/mol |
MDL Number:
MFCD00011798 CAS Number: 420-37-1 UN: 1759 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...