(S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
(S)-(-)-5,5',6,6'-Tetramethyl-3,3'-di-t-butyl-1,1'-biphenyl-2,2'-diol, (S)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, (S)-BIPHEN H2
08-2047-500MG
08-2047-100MG
08-2047-2G
100207-944EA
294.4
USD
100207-944
100207-942
100207-946
(S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol ≥99%
(S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol
BIPHEN
Formula:
C₂₄H₃₄O₂ |
MDL Number:
MFCD01862439 CAS Number: 205927-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...