(R)-(+)-5,5',6,6'-Tetramethyl-3,3'-di-t-butyl-1,1'-biphenyl-2,2'-diol ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: (R)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, (R)-BIPHEN H2

08-2046-500MG 08-2046-100MG 08-2046-2G
100207-938EA 300.06 USD
100207-938 100207-936 100207-940
(R)-(+)-5,5',6,6'-Tetramethyl-3,3'-di-t-butyl-1,1'-biphenyl-2,2'-diol ≥99%
(R)-(+)-5,5',6,6'-Tetramethyl-3,3'-di-t-butyl-1,1'-biphenyl-2,2'-diol

BIPHEN

Formula: C₂₄H₃₄O₂
MW: 354.5256 g/mol
MDL Number: MFCD01862439
CAS Number: 329735-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR