(+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
rac-BIPHEN H2, 3,3'-Di-tert-butyl-5,5',6,6'-tetramethyl-[1,1'-biphenyl]-2,2'-diol, racemic-5,5',6,6'-Tetramethyl-3,3'-di-t-butyl-1,1'-biphenyl-2,2'-diol
08-2045-5G
08-2045-25G
100208-292EA
172.68
USD
100208-292
100208-290
(+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol ≥99%
(+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol
BIPHEN
Formula:
C₂₄H₃₄O₂ Melting Pt: 163…165 °C |
MDL Number:
MFCD01862439 CAS Number: 101203-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...