(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-(+)-2,2'-Dihydroxy-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, (R)-5,5',6,6',7,7',8,8'-Octahydro[1,1'-binaphthalene]-2,2'-diol

08-2035-250MG 08-2035-1G
100209-568EA 219.38 USD
100209-568 100198-540
(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥99%
(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol
Formula: C₂₀H₂₂O₂
MW: 294.39 g/mol
MDL Number: MFCD02093485
CAS Number: 65355-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR