racemic-4-Hydroxy[2.2]paracyclophane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

08-2027-250MG 08-2027-1G
101418-444EA 65.11 USD
101418-444 101418-446
racemic-4-Hydroxy[2.2]paracyclophane ≥97%
racemic-4-Hydroxy[2.2]paracyclophane

Paracyclophane

MDL Number: MFCD00268257
CAS Number: 157018-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR