(4R,5R)-(-)-2,2-Dimethyl-α;,α;,α;',α;'-tetra(1-naphthyl)-1,3-dioxolane-4,5-dimethanol ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R,R)-1-Nph-TADDOL, ((4R,5R)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(di(naphthalen-1-yl)methanol)

08-2004-2G 08-2004-500MG
100208-620EA 530.77 USD
100208-620 100195-992
(4R,5R)-(-)-2,2-Dimethyl-α;,α;,α;',α;'-tetra(1-naphthyl)-1,3-dioxolane-4,5-dimethanol ≥97%
(4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetra(1-naphthyl)-1,3-dioxolane-4,5-dimethanol

TADDOL

Formula: C₄₇H₃₈O₄
MW: 666.8 g/mol
Melting Pt: 200 °C (dec.)
MDL Number: MFCD00192091
CAS Number: 137536-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR