(3S)-2,2'-Diphenyl[3,3'-biphenanthrene]-4,4'-diol ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
08-1706-500MG
08-1706-100MG
101418-442EA
1542.77
USD
101418-442
101418-440
(3S)-2,2'-Diphenyl[3,3'-biphenanthrene]-4,4'-diol ≥98%
(3S)-2,2'-Diphenyl[3,3'-biphenanthrene]-4,4'-diol
Formula:
C₄₀H₂₆O₂ |
MDL Number:
MFCD08459339 CAS Number: 147702-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...