(2S)-(-)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-VANOL, (2S)-3,3'-Diphenyl[2,2'-binaphthalene]-1,1'-diol

08-1702-100MG 08-1702-500MG
101418-432EA 315.63 USD
101418-432 101418-434
(2S)-(-)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol ≥98%
(2S)-(-)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol
Formula: C₃₂H₂₂O₂
MW: 438.5159 g/mol
MDL Number: MFCD08459338
CAS Number: 147702-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR