(2R)-(+)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-VANOL, (2R)-3,3'-Diphenyl[2,2'-binaphthalene]-1,1'-diol

08-1700-500MG 08-1700-100MG
101418-430EA 1245.54 USD
101418-430 101418-428
(2R)-(+)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol ≥98%
(2R)-(+)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol
Formula: C₃₂H₂₂O₂
MW: 438.5100 g/mol
MDL Number: MFCD08459338
CAS Number: 147702-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR