(S)-(-)-1,1'-Bi-2-naphthol ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol

08-1001-5G 08-1001-25G 08-1001-1G
100198-536EA 158.52 USD
100198-536 100198-534 100198-532
(S)-(-)-1,1'-Bi-2-naphthol ≥99%
(S)-(-)-1,1'-Bi-2-naphthol

BINOL

Formula: C₂₀H₁₄O₂
Melting Pt: 207…210 °C
MDL Number: MFCD00004068
CAS Number: 18531-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR