1,2-Diiodo-4,5-di-n-octyloxybenzene

Supplier: STREM CHEMICALS INC MS
Warning

08-0722-250MG 08-0722-1G
101442-216EA 205.23 USD
101442-216 101442-218
1,2-Diiodo-4,5-di-n-octyloxybenzene
1,2-Diiodo-4,5-di-n-octyloxybenzene
MDL Number: MFCD11045084
CAS Number: 248277-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR