1,2-Diiodo-4,5-di-n-butoxybenzene

Supplier: STREM CHEMICALS INC MS
Warning

08-0720-250MG 08-0720-1G
101442-212EA 209.48 USD
101442-212 101442-214
1,2-Diiodo-4,5-di-n-butoxybenzene
1,2-Diiodo-4,5-di-n-butoxybenzene
MDL Number: MFCD11040285
CAS Number: 1005340-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR