(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol

08-0650-250MG 08-0650-1G
100197-050EA 263.26 USD
100197-050 100208-282
(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥99%
(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol
Formula: C₂₀H₂₀Br₂O₂
MW: 452.1800 g/mol
MDL Number: MFCD03093977
CAS Number: 65355-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR