(S)-(-)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene, (S)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl

08-0621-1G 08-0621-250MG
100198-520EA 317.05 USD
100198-520 100209-566
(S)-(-)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl ≥98%
(S)-(-)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl
Formula: C₂₄H₂₀Br₂O₄
MW: 532.23 g/mol
Melting Pt: 133 °C
MDL Number: MFCD03788936
CAS Number: 211560-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR