(S)-(+)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(S)-6,6'-Dibromo-[1,1'-binaphthalene]-2,2'-diol
08-0606-500MG
08-0606-2G
100198-514EA
208.06
USD
100198-514
100206-254
(S)-(+)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98%
(S)-(+)-6,6'-Dibromo-1,1'-bi-2-naphthol
Formula:
C₂₀H₁₂Br₂O₂ MW: 444.12 g/mol Melting Pt: 92…96 °C |
MDL Number:
MFCD00798290 CAS Number: 80655-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...