(R)-(-)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-6,6'-Dibromo-[1,1'-binaphthalene]-2,2'-diol

08-0605-500MG 08-0605-2G
100198-512EA 208.06 USD
100198-512 100206-252
(R)-(-)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98%
(R)-(-)-6,6'-Dibromo-1,1'-bi-2-naphthol
Formula: C₂₀H₁₂Br₂O₂
MW: 444.12 g/mol
Melting Pt: 92…96 °C
MDL Number: MFCD00798290
CAS Number: 65283-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR