Dibenzo-24-crown-8-(dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane) ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Dibenzo-24-crown-8
08-0510-1G
08-0510-5G
100209-562EA
159.94
USD
100209-562
100210-784
Dibenzo-24-crown-8-(dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane) ≥98%
Dibenzo-24-crown-8-(dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane)
Formula:
C₂₄H₃₂O₈ MW: 448.51 g/mol Melting Pt: 103…104 °C |
MDL Number:
MFCD00005101 CAS Number: 14174-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...