(S)-(+)-1,1'-Binaphthyl-2,2'-diylbis(trifluoromethanesulfonate) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-(+)-1,1'-Bi-2-naphthol bis(trifluoromethanesulphonate), (S)-[1,1-Binaphthalene]-2,2-diyl bis(trifluoromethanesulphonate), (S)-(+)-1,1'-Bi-2-naphthol Bis(trifluoromethanesulphonate)

08-0161-250MG 08-0161-1G 08-0161-5G
100201-374EA 48.12 USD
100201-374 100201-376 100201-378
(S)-(+)-1,1'-Binaphthyl-2,2'-diylbis(trifluoromethanesulfonate) ≥98%
(S)-(+)-1,1'-Binaphthyl-2,2'-diylbis(trifluoromethanesulfonate)
Formula: C₂₂H₁₂F₆O₆S₂
MW: 550.44 g/mol
Melting Pt: 82…85 °C
MDL Number: MFCD00274615
CAS Number: 128544-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR