(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulfonate) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-(-)-1,1'-Binaphthol-2,2'-bis(trifluoromethanesulphonate), (R)-(-)-1,1'-Bi-2-naphthol Bis(trifluoromethanesulphonate), (R)-(-)-1,1'-Bi-2-naphthol bis(trifluoromethanesulphonate), (R)-[1,1'-Binaphthalene]-2,2'-diyl bis(trifluoromethanesulphonate)

08-0160-1G 08-0160-5G 08-0160-250MG
100201-370EA 138.71 USD
100201-370 100201-372 100201-368
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulfonate) ≥98%
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulfonate)
Formula: C₂₂H₁₂F₆O₆S₂
Melting Pt: 82…85 °C
MDL Number: MFCD00274615
CAS Number: 126613-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR