Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Benzo-15-crown-5
08-0150-1G
08-0150-5G
100201-364EA
75.02
USD
100201-364
100201-366
Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥97%
Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane)
Formula:
C₁₄H₂₀O₅ MW: 268.31 g/mol Melting Pt: 76…78 °C |
MDL Number:
MFCD00005945 CAS Number: 14098-44-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...