Tris(pyrazol-1-yl)methane ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Tris(1-pyrazolyl)methane
07-3200-1G
07-3200-250MG
100210-966EA
198.15
USD
100210-966
100197-334
Tris(pyrazol-1-yl)methane ≥98%
Tris(pyrazol-1-yl)methane
Formula:
C₁₀H₁₀N₆ MW: 214.23 g/mol Melting Pt: 103…104 °C |
MDL Number:
MFCD00040255 CAS Number: 80510-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...