1,4,7-Triazacyclononane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: 1,4,7-Triazonane, TACN

07-2500-25MG 07-2500-100MG
100204-816EA 49.54 USD
100204-816 100209-558
1,4,7-Triazacyclononane ≥97%
1,4,7-Triazacyclononane
Formula: C₆H₁₅N₃
MW: 129.2 g/mol
Flash Pt: 230 °F
MDL Number: MFCD00012358
CAS Number: 4730-54-5
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR