Tris(2-aminoethyl)amine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
N1,N1-Bis(2-aminoethyl)ethane-1,2-diamine, TREN, 2,2',2''-Triaminotriethylamine
07-2400-10G
07-2400-50G
100201-362EA
53.78
USD
100201-362
100202-874
Tris(2-aminoethyl)amine ≥98%
Tris(2-aminoethyl)amine
Formula:
C₆H₁₈N₄ MW: 146.24 g/mol Boiling Pt: 98…99 °C (1 mmHg) Density: 0,977 Flash Pt: >230 °F |
MDL Number:
MFCD00008177 CAS Number: 4097-89-6 UN: 2927 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...