N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (1R,2R)-(-)-N-(4-toluenesulphonyl)-1,2-diphenylethylenediamine, (R,R)-TsDPEN, (1R,2R)-(-)-N-(4-Toluenesulphonyl)-1,2-diphenylethylenediamine

07-2371-10G 07-2371-500MG 07-2371-2G
101442-160EA 598.71 USD
101442-160 100201-360 100206-250
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide ≥98%
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide

CATHy Catalyst, DPEN

Formula: C₂₁H₂₂N₂O₂S
MW: 366.48 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD02093428
CAS Number: 144222-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR