Tetrasodium meso-tetra(sulfonatophenyl)porphine dodecahydrate ≥95%

Supplier: STREM CHEMICALS INC MS

07-2340-250MG 07-2340-1G
100198-078EA 232.12 USD
100198-078 100198-080
Tetrasodium meso-tetra(sulfonatophenyl)porphine dodecahydrate ≥95%
Formula: C₄₄H₂₆N₄Na₄O₁₂S₄·12H₂O
MDL Number: MFCD00151011
CAS Number: 122451-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR