5,10,15,20-Tetra(4-pyridyl)porphyrin 97

Supplier: STREM CHEMICALS INC MS

Synonyms: 5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine, meso-Tetra(4-pyridyl)porphine

07-2300-5G 07-2300-1G
100209-244EA 758.65 USD
100209-244 100204-484
5,10,15,20-Tetra(4-pyridyl)porphyrin 97
5,10,15,20-Tetra(4-pyridyl)porphyrin

Porphine

Formula: C₄₀H₂₆N₈
MW: 618.69 g/mol
MDL Number: MFCD00009626
CAS Number: 16834-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR