meso-Tetra(4-carboxyphenyl)porphine ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Tetrakis(4-carboxyphenyl)porphyrin, 4,4,4,4-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid), TCPP, Tetrakis (4-carboxyphenyl) porphyrin

07-1970-250MG 07-1970-1G
100210-964EA 100.49 USD
100210-964 100199-726
meso-Tetra(4-carboxyphenyl)porphine ≥98%
meso-Tetra(4-carboxyphenyl)porphine

Porphine

Formula: C₄₈H₃₀N₄O₈
MW: 790.79 g/mol
MDL Number: MFCD00064860
CAS Number: 14609-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR