N,N'-Bis(2-aminoethyl)-1,3-propanediamine ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: 3,7-Diaza-1,9-nonanediamine, N1,N1'-(Propane-1,3-diyl)bis(ethane-1,2-diamine), 1,4,8,11-Tetraazaundecane

07-1965-5G 07-1965-1G 07-1965-25G
100195-988EA 234.95 USD
100195-988 100201-354 100195-986
N,N'-Bis(2-aminoethyl)-1,3-propanediamine ≥97%
N,N'-Bis(2-aminoethyl)-1,3-propanediamine
Formula: C₇H₂₀N₄
MW: 160.27 g/mol
Boiling Pt: 142…145 °C (8 mmHg)
Density: 0,96
Flash Pt: 230 °F
MDL Number: MFCD00008174
CAS Number: 4741-99-5
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR