N,N'-Bis(3-aminopropyl)propane-1,3-diamine ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: N,N'-Bis(3-aminopropyl)-1,3-propanediamine, 1,5,9,13-Tetraazatridecane

07-1963-25G 07-1963-5G
100207-230EA 1062.95 USD
100207-230 100199-328
N,N'-Bis(3-aminopropyl)propane-1,3-diamine ≥97%
N,N'-Bis(3-aminopropyl)propane-1,3-diamine
Formula: C₉H₂₄N₄
MW: 188.31 g/mol
Boiling Pt: 98…103 °C (1 mmHg)
Density: 0,92
Flash Pt: 230 °F
MDL Number: MFCD00008213
CAS Number: 4605-14-5
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR